Abnova
Manufacturer Code:H00002875M02A
Catalog # 89-001-457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F10 |
Immunogen | GPT (AAH18207.1 1 a.a. ~ 496 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant GPT.
For Research Use Only
Protein Aliases: AAT1; alanine aminotransferase; Alanine aminotransferase 1; ALT1; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1; Glutamic--pyruvic transaminase 1; glutamic-alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); GPT 1; GPT1
Gene Aliases: AAT1; ALT1; GPT; GPT1
UniProt ID: (Human) P24298
Entrez Gene ID: (Human) 2875
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.