Novus Biologicals
Manufacturer Code:NB12011318
Catalog # NB12011318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: EEFATEGTDRKDVFFY-K corresponding to amino acids 38-53 of Human gamma Tubulin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gamma-1-tubulin; gamma-1-tubulin Gamma-tubulin complex component 1 GCP-1 TUBG TUBGCP1 tubulin gamma-1 chain tubulin gamma 1 tubulin gamma polypeptide; Gamma-tubulin complex component 1; GCP-1; Tubulin gamma-1 chain; tubulin, gamma 1; tubulin, gamma polypeptide
Gene Aliases: CDCBM4; GCP-1; TUBG; TUBG1; TUBGCP1
UniProt ID: (Human) P23258
Entrez Gene ID: (Human) 7283
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.