Proteintech
Manufacturer Code:151761AP150UL
Catalog # 50-100-2590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | gamma Tubulin Fusion Protein Ag7070 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gamma 1 tubulin gamma Tubulin GCP 1 TUBG TUBG1 TUBGCP1 Tubulin gamma 1 chain tubulin gamma 1; gamma tubulin; Gamma-1-tubulin; Gamma-tubulin complex component 1; GCP-1; Tubulin gamma-1 chain; tubulin, gamma 1; tubulin, gamma polypeptide
Gene Aliases: 1500010O08Rik; AI451582; AI503389; CDCBM4; GCP-1; TUBG; TUBG1; TUBGCP1
UniProt ID: (Human) P23258, (Dog) Q9GKK5, (Mouse) P83887, (Rat) P83888
Entrez Gene ID: (Human) 7283, (Dog) 403695, (Mouse) 103733, (Rat) 252921
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.