Novus Biologicals
Manufacturer Code:NB10074435
Catalog # NB10074435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F2 |
Immunogen | This gamma H2AX [p Ser139] Antibody (3F2) was developed against a synthetic peptide sequence surrounding phosphorylated Ser139. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H2A histone family member X H2A.X H2a/x H2AX histone H2AXhistone H2A.x YH2AX; H2A histone family, member X; H2a/x; H2AX histone; Histone H2A.X; Histone H2AX
Gene Aliases: H2A.X; H2A/X; H2AFX; H2AX
UniProt ID: (Human) P16104
Entrez Gene ID: (Human) 3014
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.