Invitrogen
Manufacturer Code:PA532295
Catalog # PIPA532295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from the C-terminus of human catenin-gamma protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARVD12 CTNNG DP3 DPIII PDGB PKGB catenin cadherin-associated protein gamma 80kD catenin cadherin-associated protein gamma 80kDa catenin gamma desmoplakin III desmoplakin-3 gamma-catenin; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; Desmoplakin III; Desmoplakin-3; Junction plakoglobin
Gene Aliases: ARVD12; CTNNG; DP3; DPIII; JUP; PDGB; PKGB
UniProt ID: (Human) P14923
Entrez Gene ID: (Human) 3728
Molecular Function: cell adhesion molecule cytoskeletal protein signaling molecule storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.