Invitrogen
Manufacturer Code:MA537896
Catalog # PIMA537896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2440 |
Immunogen | Recombinant protein of human AP1G1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AA409002 adapter-related protein complex 1 subunit gamma-1 adaptor protein complex AP-1 gamma 1 subunit Adaptor protein complex AP-1 subunit gamma-1 adaptor protein complex AP-1 gamma 1 subunit adaptor related protein complex 1 gamma 1 subunit adaptor related protein complex 1 subunit gamma 1 adaptor-related protein complex 1 gamma 1 subunit adaptor-related protein complex 1 subunit gamma-1 adaptor-related protein complex 1 gamma 1 subunit ADTG AP-1 complex subunit gamma-1 AP1G1 AU041323 AW551707 CLAPG1 clathrin assembly protein complex 1 gamma large chain clathrin assembly protein complex 1 gamma-1 large chain clathrin-associated/assembly/adaptor protein large gamma 1 D8Ertd374e gamma adaptin gamma1-adaptin gamma-adaptin golgi adaptor HA1/AP1 adaptin gamma subunit golgi adaptor HA1/AP1 adaptin subunit gamma-1 MGC18255 testicular tissue protein Li 21; adapter-related protein complex 1 subunit gamma-1; adaptor protein complex AP-1 gamma 1 subunit; Adaptor protein complex AP-1 subunit gamma-1; adaptor protein complex AP-1, gamma 1 subunit; adaptor-related protein complex 1 gamma 1 subunit; Adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; AP-1 complex subunit gamma-1; clathrin assembly protein complex 1 gamma large chain; Clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; gamma adaptin; Gamma-adaptin; Gamma1-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; Golgi adaptor HA1/AP1 adaptin subunit gamma-1; testicular tissue protein Li 21
Gene Aliases: AA409002; ADTG; AP1G1; AU041323; AW551707; CLAPG1; D8Ertd374e
UniProt ID: (Human) O43747, (Mouse) P22892
Entrez Gene ID: (Human) 164, (Mouse) 11765, (Rat) 171494
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.