Invitrogen
Manufacturer Code:PA529965
Catalog # PIPA529965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 20 and 299 of Human gamma Adaptin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 1 subunit gamma-1; Adaptor protein complex AP-1 subunit gamma-1; adaptor-related protein complex 1 gamma 1 subunit; Adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; ADTG CLAPG1 AP-1 complex subunit gamma-1 adapter-related protein complex 1 subunit gamma-1 adaptor protein complex AP-1 subunit gamma-1 clathrin assembly protein complex 1 gamma large chain clathrin assembly protein complex 1 gamma-1 large chain clathrin-associated/a; AP-1 complex subunit gamma-1; clathrin assembly protein complex 1 gamma large chain; Clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; gamma adaptin; Gamma1-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; Golgi adaptor HA1/AP1 adaptin subunit gamma-1; testicular tissue protein Li 21
Gene Aliases: ADTG; AP1G1; CLAPG1
UniProt ID: (Human) O43747
Entrez Gene ID: (Human) 164
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.