Abnova
Manufacturer Code:H00003170D01P
Catalog # 89-017-724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FOXA2 (NP_068556.1 1 a.a. ~ 457 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human FOXA2 protein.
For Research Use Only
Protein Aliases: Forkhead box protein A2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3, beta; Hepatocyte nuclear factor 3-beta; HNF-3-beta; HNF-3B; HNF3B; MGC19807; TCF-3B; TCF3B; Transcription factor 3B
Gene Aliases: FOXA2; HNF3B; TCF3B
UniProt ID: (Human) Q9Y261
Entrez Gene ID: (Human) 3170
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.