Abnova
Manufacturer Code:H00002246D01P
Catalog # 89-017-633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FGF1 (NP_000791.1 1 a.a. ~ 155 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human FGF1 protein.
For Research Use Only
Protein Aliases: Acidic fibroblast growth factor; AFGF; beta-endothelial cell growth factor; ECGF; ECGF-beta; ECGFA; ECGFB; Endothelial cell growth factor; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; FGF-1; FGF-alpha; FGFA; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); GLIO703; HBGF-1; HBGF1; Heparin-binding growth factor 1
Gene Aliases: AFGF; ECGF; ECGF-beta; ECGFA; ECGFB; FGF-1; FGF-alpha; FGF1; FGFA; GLIO703; HBGF-1; HBGF1
UniProt ID: (Human) P05230
Entrez Gene ID: (Human) 2246
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.