Abnova
Manufacturer Code:H00002495B03
Catalog # 89-005-399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | FTH1 (NP_002023 1 a.a. ~ 183 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human FTH1 protein.
For Research Use Only
Protein Aliases: apoferritin; Cell proliferation-inducing gene 15 protein; Ferritin H subunit; Ferritin heavy chain; Ferritin heavy chain, N-terminally processed; ferritin, heavy polypeptide 1; FHC; FTH; FTHL6; MGC104426; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15
Gene Aliases: FHC; FTH; FTH1; FTHL6; HFE5; OK/SW-cl.84; PIG15; PLIF
UniProt ID: (Human) P02794
Entrez Gene ID: (Human) 2495
Molecular Function:
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.