Abnova
Manufacturer Code:H00051010D01P
Catalog # 89-018-469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EXOSC3 (NP_057126.2 1 a.a. ~ 275 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human EXOSC3 protein.
For Research Use Only
Protein Aliases: bA3J10.7; CGI-102; Exosome complex component RRP40; exosome complex exonuclease RRP40; Exosome component 3; hRrp40p; MGC15120; MGC723; p10; Ribosomal RNA-processing protein 40; RP11-3J10.8; RRP40; Rrp40p
Gene Aliases: bA3J10.7; CGI-102; EXOSC3; hRrp-40; p10; PCH1B; RRP40; Rrp40p
UniProt ID: (Human) Q9NQT5
Entrez Gene ID: (Human) 51010
Molecular Function: RNA binding protein esterase exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.