Invitrogen
Manufacturer Code:PA569515
Catalog # PIPA569515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of fruit fly exd |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anon-fast-evolving-1H3; CG8933-PA; CG8933-PB; CG8933-PD; CG8933-PE; Dpbx; exd CG8933; exd-PA; exd-PB; exd-PD; exd-PE; Homeobox protein extradenticle; LD03509p; Pbx1-oncogene-like
Gene Aliases: anon-EST:fe1H3; CG8933; DExd; Dm-EXD; Dmel\CG8933; Dmel_CG8933; Dpbx; EXD; l(1)IV; lincRNA.S9404; Pbx1; td48
UniProt ID: (Fruit fly) P40427
Entrez Gene ID: (Fruit fly) 32567
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.