Abnova
Manufacturer Code:H00002068B01P
Catalog # 89-016-784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ERCC2 (NP_000391.1 1 a.a. ~ 760 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ERCC2 protein.
For Research Use Only
Protein Aliases: Basic transcription factor 2 80 kDa subunit; BTF2 p80; COFS2; CXPD; DNA excision repair protein ERCC-2; DNA repair protein complementing XP-D cells; EM9; excision repair cross-complementation group 2; excision repair cross-complementing rodent repair deficiency, complementation group 2; General transcription and DNA repair factor IIH helicase subunit XPD; MGC102762; MGC126218; MGC126219; TFIIH 80 kDa subunit; TFIIH basal transcription factor complex 80 kDa subunit; TFIIH basal transcription factor complex helicase subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH p80; TFIIH subunit XPD; TTD; xeroderma pigmentosum complementary group D; Xeroderma pigmentosum group D-complementing protein; XPD
Gene Aliases: COFS2; EM9; ERCC2; TFIIH; TTD; TTD1; XPD; XPDC
UniProt ID: (Human) P18074
Entrez Gene ID: (Human) 2068
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.