Invitrogen
Manufacturer Code:PA569510
Catalog # PIPA569510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of fruit fly eve |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG2328-PA; complementation group F; eve CG2328; eve PRE/TRE element; eve-PA; even-skiped; even-skipped; evenskip; evenskipped; group V; group VI; lethal(2)46Ce; Segmentation protein even-skipped
Gene Aliases: 10.5; 10.9; 14.10; 20.35; CG2328; dm-eve; Dmel\CG2328; Dmel_CG2328; E(eve); EVE; eve2; even; F; l(2)46Ce; l(2)46CFg; l(2)46CFh; l(2)46CFj; l(2)46CFp; l(2)46Cg; V; VI
UniProt ID: (Fruit fly) P06602
Entrez Gene ID: (Fruit fly) 36039
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.