Novus Biologicals
Manufacturer Code:NBP233918
Catalog # NBP233918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: EFIQGEALDPKHVCNPAIFRLIARQLAKIHAIHAHNGWIPKSNLWLKMGKYFSLIPTGFADEDINKRFLSDIPSSQILQEEMTWMKEILSNLGSPVVLCHNDLLCKNIIYNE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.82 EKI EKI 1 EKI1putative protein product of Nbla10396 ethanolamine kinase 1 Nbla10396; EKI 1; Ethanolamine kinase 1; putative protein product of Nbla10396
Gene Aliases: EKI; EKI 1; EKI1; ETNK1; Nbla10396
UniProt ID: (Human) Q9HBU6
Entrez Gene ID: (Human) 55500
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.