Novus Biologicals
Manufacturer Code:NBP303768100UL
Catalog # NB093680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 585-854 of human espin (NP_113663.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Autosomal recessive deafness type 36 protein; autosomal recessive deafness type 36 protein DKFZp434A196 DKFZp434G2126 espin; Ectoplasmic specialization protein; Espin
Gene Aliases: DFNB36; ESPN; LP2654
UniProt ID: (Human) B1AK53
Entrez Gene ID: (Human) 83715
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.