Abnova
Manufacturer Code:H00009583B01
Catalog # 89-006-810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENTPD4 (AAH34477.1 1 a.a. ~ 571 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ENTPD4 protein.
For Research Use Only
Protein Aliases: Ectonucleoside triphosphate diphosphohydrolase 4; golgi luminal UDPase; Golgi UDPase; guanosine-diphosphatase like protein; KIAA0392; LALP70; LAP70; LYSAL1; lysosomal apyrase-like 1; Lysosomal apyrase-like protein of 70 kDa; NTPDase 4; NTPDase-4; UDPase; Uridine-diphosphatase
Gene Aliases: ENTPD4; KIAA0392; LALP70; LAP70; LYSAL1; NTPDase-4; UDPase
UniProt ID: (Human) Q9Y227
Entrez Gene ID: (Human) 9583
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.