Invitrogen
Manufacturer Code:PA557982
Catalog # PIPA557982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human eRF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D5S1995 ERF ERF1 RF1 SUP45L1 TB3-1 eukaryotic peptide chain release factor subunit 1 eukaryotic release factor 1 polypeptide chain release factor 1 protein Cl1 sup45 (yeast omnipotent suppressor 45) homolog-like 1; Eukaryotic peptide chain release factor subunit 1; Eukaryotic release factor 1; polypeptide chain release factor 1; Protein Cl1; sup45 (yeast omnipotent suppressor 45) homolog-like 1; TB3-1
Gene Aliases: D5S1995; ERF; ERF1; ETF1; RF1; SUP45L1; TB3-1
UniProt ID: (Human) D3DQC1
Entrez Gene ID: (Human) 2107
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.