Novus Biologicals
Manufacturer Code:NB300500IR
Catalog # NB300500IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues S(1179) LQERQLRGAVPWAFD(1194) of human eNOS. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: cNOS; Constitutive NOS; Constitutive NOS EC 1.14.13.39 ECNOS endothelial Endothelial NOS eNOScNOS nitric oxide synthase 3 (endothelial cell) NOS type III NOSIII; EC-NOS; Endothelial NOS; eNOS; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; Nitric oxide synthase, endothelial; NOS type III; NOSIII
Gene Aliases: ECNOS; eNOS; NOS3
UniProt ID: (Human) P29474
Entrez Gene ID: (Human) 4846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.