Invitrogen
Manufacturer Code:PA5102489
Catalog # PIPA5102489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EIF6 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110004P11 AA408895 b(2)gcn B(2)GCN homolog B4 integrin interactor CAB EIF3A Eif6 eIF-6 eIF-6 {ECO:0000255 eukaryotic translation initiation factor 3A eukaryotic translation initiation factor 6 eukaryotic translation initiation factor 6 {ECO:0000255 HAMAP-Rule:MF_03132} imc-415 integrin beta 4 binding protein Itgb4bp OK/SW-cl.27 p27 beta-4 integrin-binding protein p27(BBP) p27BBP; B(2)GCN homolog; B4 integrin interactor; CAB; eIF-6; eIF-6 {ECO:0000255|HAMAP-Rule:MF_03132}; EIF6; eukaryotic translation initiation factor 3A; Eukaryotic translation initiation factor 6; eukaryotic translation initiation factor 6 {ECO:0000255|HAMAP-Rule:MF_03132}; integrin beta 4 binding protein; p27 beta-4 integrin-binding protein; p27(BBP)
Gene Aliases: 1110004P11; AA408895; b(2)gcn; CAB; eIF-6; EIF3A; EIF6; imc-415; ITGB4BP; OK/SW-cl.27; p27(BBP); p27BBP
UniProt ID: (Human) P56537, (Rat) Q3KRD8, (Mouse) O55135
Entrez Gene ID: (Human) 3692, (Rat) 305506, (Mouse) 16418
Molecular Function: RNA binding protein nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.