Novus Biologicals
Manufacturer Code:NB200133
Catalog # NB200133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide representing the C-terminus of the Human EIF5 A2 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-5A-2; eIF-5A-2 EIF-5A2 eIF5AII eukaryotic initiation factor 5A Eukaryotic initiation factor 5A isoform 2 eukaryotic translation initiation factor 5A2 eukaryotic translation initiation factor 5A-2; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; Eukaryotic translation initiation factor 5A-2
Gene Aliases: EIF-5A2; EIF5A2; eIF5AII
UniProt ID: (Human) Q9GZV4
Entrez Gene ID: (Human) 56648
Molecular Function: RNA binding protein nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.