Invitrogen
Manufacturer Code:MA535512
Catalog # PIMA535512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0999 |
Immunogen | A synthesized peptide derived from human eIF5A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AA410058 D19Wsu54e Eif4d eIF-4D EIF5A eIF-5A EIF5A1 eIF-5A1 eIF-5A-1 eIF5AI Eukaryotic initiation factor 5A isoform 1 Eukaryotic translation initiation factor 5A-1 MGC104255 MGC99547 OTTHUMP00000128384 Rev-binding factor uORF A; eIF-4D; eIF-5A; eIF-5A-1; eIF-5A1; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; Eukaryotic translation initiation factor 5A-1; Rev-binding factor
Gene Aliases: AA410058; D19Wsu54e; eIF-4D; EIF-5A; eIF-5A-1; eIF-5A1; Eif4d; EIF5A; EIF5A1; eIF5AI
UniProt ID: (Human) P63241, (Mouse) P63242, (Rat) Q3T1J1
Entrez Gene ID: (Human) 1984, (Mouse) 276770, (Rat) 287444
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.