Novus Biologicals
Manufacturer Code:NBP297002G
Catalog # NB112253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant eIF5A (NP_001961.1 Met1-Lys154). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-4D; eIF-4D eIF-5A eIF-5A1 eIF-5A-1 EIF5A1 eIF5AI eukaryotic initiation factor 5A Eukaryotic initiation factor 5A isoform 1 eukaryotic translation initiation factor 5A eukaryotic translation initiation factor 5A-1 MGC104255 MGC99547 Rev-binding factor; eIF-5A; eIF-5A-1; eIF-5A1; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; Eukaryotic translation initiation factor 5A-1; Rev-binding factor
Gene Aliases: EIF-5A; EIF5A; EIF5A1; eIF5AI
UniProt ID: (Human) P63241
Entrez Gene ID: (Human) 1984
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.