R&D Systems
Manufacturer Code:AF7558
Catalog # AF7558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human eIF5A Lys85-Lys154 Accession # P63241 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-4D; eIF-5A; eIF-5A-1; eIF-5A1; eIF4D eIF-4D eIF-5A eIF-5A1 eIF-5A-1 EIF5A1 eIF5AI eukaryotic initiation factor 5A Eukaryotic initiation factor 5A isoform 1 eukaryotic translation initiation factor 5A eukaryotic translation initiation factor 5A-1 MGC104255 MGC99547 Rev-binding factor; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; Eukaryotic translation initiation factor 5A-1; Rev-binding factor
Gene Aliases: EIF-5A; EIF5A; EIF5A1; eIF5AI
UniProt ID: (Human) P63241
Entrez Gene ID: (Human) 1984
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.