Invitrogen
Manufacturer Code:PA583149
Catalog # PIPA583149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human EIF4H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-4H; Eukaryotic translation initiation factor 4H; KIAA0038 WBSCR1 WSCR1; Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosomal region 1 protein homolog; Williams-Beuren syndrome chromosome region 1; Williams-Beuren syndrome chromosome region 1 homolog
Gene Aliases: AU018978; D5Ertd355e; E430026L18Rik; Ef4h; eIF-4H; EIF4H; KIAA0038; mKIAA0038; WBSCR1; WSCR1
UniProt ID: (Human) Q15056, (Mouse) Q9WUK2, (Rat) Q5XI72
Entrez Gene ID: (Human) 7458, (Mouse) 22384, (Rat) 288599
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.