Novus Biologicals
Manufacturer Code:NBP15724820UL
Catalog # NBP15724820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to WBSCR1 The peptide sequence was selected from the middle region of WBSCR1. Peptide sequence DSRDDFNSGFRDDFLGGRGGSRPGDRRTGPPMGSRFRDGPPLRGSNMDFR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-4H; Eukaryotic translation initiation factor 4H; eukaryotic translation initiation factor 4H KIAA0038eIF-4H WBSCR1 Williams-Beuren syndrome chromosome region 1 WSCR1Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosome region 1
Gene Aliases: eIF-4H; EIF4H; KIAA0038; WBSCR1; WSCR1
UniProt ID: (Human) Q15056
Entrez Gene ID: (Human) 7458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.