Novus Biologicals
Manufacturer Code:NBP216309
Catalog # NBP216309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human EIF4G3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-4-gamma 3; eIF-4-gamma II; eIF-4G 3 eIF-4-gamma 3 eIF-4-gamma II eIF4GIIeIF4G 3 eukaryotic translation initiation factor 4 gamma 3 eukaryotic translation initiation factor 4 gamma 3; eIF4GII; Eukaryotic translation initiation factor 4 gamma 3
Gene Aliases: eIF-4G 3; eIF4G 3; EIF4G3; eIF4GII
UniProt ID: (Human) O43432
Entrez Gene ID: (Human) 8672
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.