Invitrogen
Manufacturer Code:MA525413
Catalog # PIMA525413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F2 |
Immunogen | Full length human recombit protein of EIF4E2 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF-4E type 2; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; Eukaryotic translation initiation factor 4E homologous protein; Eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E type 2 eIF4E type 2 eIF-4E type 2 mRNA cap-binding protein type 3 Eukaryotic translation initiation factor 4E-like 3 Eukaryotic translation initiation factor 4E homologous protein mRNA cap-binding protein 4EHP eIF4E-like protein 4E-LP EIF4E2 EIF4EL3; Eukaryotic translation initiation factor 4E-like 3; h4EHP; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 3
Gene Aliases: 4E-LP; 4EHP; EIF4E2; EIF4EL3; IF4e
UniProt ID: (Human) O60573
Entrez Gene ID: (Human) 9470
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.