Invitrogen
Manufacturer Code:PA541279
Catalog # PIPA541279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DDX48 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-3; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; eIF-4A-III; eIF4A-III; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-III-like protein; Eukaryotic initiation factor 4A-like NUK-34; eukaryotic initiation factor 4AIII; eukaryotic translation initiation factor 4A; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 3; Eukaryotic translation initiation factor 4A3 DDX48 NUK34 NMP265 eIF4AIII; hNMP 265; NMP 265; Nuclear matrix protein 265
Gene Aliases: 2400003O03Rik; DDX48; eIF4A-III; EIF4A3; eIF4AIII; KIAA0111; mKIAA0111; MUK34; NMP265; NUK34; RCPS; zgc:56139
UniProt ID: (Human) P38919, (Bovine) Q2NL22, (Mouse) Q91VC3, (Rat) Q3B8Q2, (Zebrafish) Q7ZVA6
Entrez Gene ID: (Human) 9775, (Horse) 100056586, (Dog) 475922, (Bovine) 515145, (Guinea pig) 100728310, (Mouse) 192170, (Rat) 688288, (Zebrafish) 394053
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.