Invitrogen
Manufacturer Code:PA523264
Catalog # PIPA523264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (69 KGYDVIAQAQSGTGKTAT 86) of eIF4A2 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase eIF4A-2; BM-010 DDX2B EIF4A EIF4F eIF-4A-II eIF4A-II ATP-dependent RNA helicase eIF4A-2 eukaryotic initiation factor 4A-II eukaryotic translation initiation factor 4A; eIF-4A-II; eIF4A-II; Eukaryotic initiation factor 4A-II; eukaryotic translation initiation factor 4A; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A1; protein synthesis initiation factor 4AII; translational eukaryotic inititation factor 4AII
Gene Aliases: 4833432N07Rik; BM-010; DDX2B; eIF-4A-II; Eif4; EIF4A; eIF4A-II; EIF4A1; EIF4A2; EIF4AII; EIF4F; RCJMB04_14a6
UniProt ID: (Human) Q14240, (Chicken) Q8JFP1, (Mouse) P10630
Entrez Gene ID: (Human) 1974, (Chicken) 395232, (Dog) 488118, (Mouse) 13682
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.