Invitrogen
Manufacturer Code:PA5112372
Catalog # PIPA5112372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Eukaryotic initiation factor 4A-II protein (1-407AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4833432N07Rik ATP-dependent RNA helicase eIF4A-2 BM-010 Ddx2b Eif4 EIF4A Eif4a2 EIF4AII eIF-4A-II eIF4A-II EIF4F Eukaryotic initiation factor 4A-II eukaryotic translation initiation factor 4A isoform 2 eukaryotic translation initiation factor 4A2 protein synthesis initiation factor 4AII RCJMB04_14a6 translational eukaryotic inititation factor 4AII; ATP-dependent RNA helicase eIF4A-2; eIF-4A-II; Eukaryotic initiation factor 4A-II
Gene Aliases: BM-010; DDX2B; eIF-4A-II; EIF4A; eIF4A-II; EIF4A2; EIF4F
UniProt ID: (Human) Q14240
Entrez Gene ID: (Human) 1974
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.