Novus Biologicals
Manufacturer Code:NBP157490
Catalog # NBP157490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to EIF4A2 (eukaryotic translation initiation factor 4A isoform 2) The peptide sequence was selected from the N terminal of EIF4A2. Peptide sequence MSGGSADYNREHGGPEGMDPDGVIESNWNEIVDNFDDMNLKESLLRGIYA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase eIF4A-2; ATP-dependent RNA helicase eIF4A-2 BM-010 DDX2Beukaryotic translation initiation factor 4A isoform 2 EC 3.6.1 EC 3.6.4.13 EIF4A eIF-4A-II eIF4A-II EIF4F eukaryotic initiation factor 4A-II eukaryotic translation initiation factor 4A eukaryotic translation initiation factor 4A2; eIF-4A-II; Eukaryotic initiation factor 4A-II
Gene Aliases: BM-010; DDX2B; eIF-4A-II; EIF4A; eIF4A-II; EIF4A2; EIF4F
UniProt ID: (Human) Q14240
Entrez Gene ID: (Human) 1974
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.