Invitrogen
Manufacturer Code:MA518796
Catalog # PIMA518796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D10 |
Immunogen | EIF2D (NP&lowbar008824.2 485 a.a. &bsim 584 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF2d; Eukaryotic translation initiation factor 2D; Eukaryotic translation initiation factor 2D HCA56 LGTN; Hepatocellular carcinoma-associated antigen 56; Ligatin
Gene Aliases: EIF2D; HCA56; LGTN
UniProt ID: (Human) P41214
Entrez Gene ID: (Human) 1939
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.