Invitrogen
Manufacturer Code:MA1078
Catalog # PIMA1078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H3 |
Immunogen | Full length human eIF2B3 produced in HEK293T cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EIF-2B EIF2Bgamma; eIF-2B GDP-GTP exchange factor subunit gamma; eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD); eukaryotic translation initiation factor 2B, subunit 3 gamma; eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa; Translation initiation factor eIF-2B subunit gamma
Gene Aliases: EIF-2B; EIF2B3; Eif2bg; EIF2Bgamma
UniProt ID: (Human) Q9NR50, (Rat) P70541
Entrez Gene ID: (Human) 8891, (Rat) 171145
Molecular Function:
G-protein modulator
RNA binding protein
enzyme modulator
guanyl-nucleotide exchange factor
nucleic acid binding
nucleotidyltransferase
transferase
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.