Invitrogen
Manufacturer Code:PA545822
Catalog # PIPA545822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human EIF2B3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-2B GDP-GTP exchange factor subunit gamma; eukaryotic translation initiation factor 2B EIF-2B EIF2Bgamma; eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD); eukaryotic translation initiation factor 2B, subunit 3 gamma; eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa; Translation initiation factor eIF-2B subunit gamma; Translation initiation factor eIF-2B subunit gamma-like protein
Gene Aliases: 1190002P15Rik; EIF-2B; EIF2B3; Eif2bg; EIF2Bgamma
UniProt ID: (Human) Q9NR50, (Rat) P70541
Entrez Gene ID: (Bovine) 534063, (Horse) 100065686, (Dog) 475379, (Human) 8891, (Rabbit) 100348406, (Guinea pig) 100723312, (Rat) 171145, (Mouse) 108067
Molecular Function: G-protein modulator RNA binding protein enzyme modulator guanyl-nucleotide exchange factor nucleic acid binding nucleotidyltransferase transferase translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.