Novus Biologicals
Manufacturer Code:NBP267353
Catalog # NBP267353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SZ01-06 |
Immunogen | Synthetic phospho-peptide surrounding S51 of eIF-2a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: 65 kDa eukaryotic translation initiation factor 2A; EIF-2 EIF-2A EIF2AEIF2 EIF-2alpha eukaryotic translation initiation factor 2 subunit 1 alpha 35kDa subunit 1 (alpha 35kD ); eIF-2A; Eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed
Gene Aliases: CDA02; EIF-2A; EIF2A; MST089; MSTP004; MSTP089
UniProt ID: (Human) Q9BY44
Entrez Gene ID: (Human) 83939
Molecular Function: RNA binding protein nuclease nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.