Invitrogen
Manufacturer Code:PA588038
Catalog # PIPA588038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-113 of human EIF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A121; eIF1; Eukaryotic translation initiation factor 1; Protein translation factor SUI1 homolog; Sui1iso1; suppressor of initiator codon mutations, related sequence 1; suppressor of initiator codon mutations-Yeast homolog related sequence 1
Gene Aliases: A121; EIF-1; EIF1; EIF1A; ISO1; SUI1; Sui1-rs1
UniProt ID: (Human) P41567, (Mouse) P48024
Entrez Gene ID: (Human) 10209, (Mouse) 20918
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.