Invitrogen
Manufacturer Code:PA5116352
Catalog # PIPA5116352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 100-133 amino acids from the C-terminal region of human EIF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A121; A121 Eif1 EIF-1 eIF13 EIF1A Eukaryotic translation initiation factor 1 ISO1 Protein translation factor SUI1 homolog SUI1 Sui1iso1 Sui1-rs1 suppressor of initiator codon mutations related sequence 1 suppressor of initiator codon mutations-Yeast homolog related sequence 1; eIF1; Eukaryotic translation initiation factor 1; Protein translation factor SUI1 homolog; Sui1iso1; suppressor of initiator codon mutations, related sequence 1; suppressor of initiator codon mutations-Yeast homolog related sequence 1
Gene Aliases: A121; EIF-1; EIF1; EIF1A; ISO1; SUI1; Sui1-rs1
UniProt ID: (Human) P41567, (Mouse) P48024
Entrez Gene ID: (Human) 10209, (Mouse) 20918
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.