Invitrogen
Manufacturer Code:PA517794
Catalog # PIPA517794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues at the amino-terminus of human eEF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EF-2; EF-2 elongation factor 2 polypeptidyl-tRNA translocase; Elongation factor 2; elongation factor-2; polypeptidyl-tRNA translocase
Gene Aliases: EEF-2; EEF2; EF-2; EF2; I79_013848; SCA26
UniProt ID: (Human) P13639, (Chicken) Q90705, (Mouse) P58252, (Rat) P05197, (Chinese hamster) P09445
Entrez Gene ID: (Human) 1938, (Chicken) 396325, (Mouse) 13629, (Rat) 29565, (Chinese hamster) 100689051
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.