Novus Biologicals
Manufacturer Code:NBP299359H
Catalog # NB084975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human eEF1A1. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCS3 CCS-3 cervical cancer suppressor 3 CTCL tumor antigen EE1A1 EEF-1 EEF1A eEF1A-1 EF1AHNGC:16303 EF1a-like protein EF-1-alpha-1 EF-Tu elongation factor 1 alpha subunit elongation factor 1-alpha 1 Elongation factor Tu Eukaryotic elongation factor 1 A-1 eukaryotic translation elongation factor 1 alpha 1 eukaryotic translation elongation factor 1 alpha 1-like 14 FLJ25721 glucocorticoid receptor AF-1 specific elongation factor GRAF-1EF LENG7 leukocyte receptor cluster (LRC) member 7 Leukocyte receptor cluster member 7 MGC102687 MGC131894 MGC16224 prostate tumor-inducing protein 1 PTI1 translation elongation factor 1 alpha 1-like 14; cervical cancer suppressor 3; CTCL tumor antigen; eEF1A-1; EF-1-alpha-1; EF-Tu; EF1a-like protein; elongation factor 1 alpha subunit; Elongation factor 1-alpha 1; Elongation factor Tu; Eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1-like 14; glucocorticoid receptor AF-1 specific elongation factor; leukocyte receptor cluster (LRC) member 7; Leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; translation elongation factor 1 alpha 1-like 14
Gene Aliases: CCS-3; CCS3; EE1A1; EEF-1; EEF1A; eEF1A-1; EEF1A1; EF-Tu; EF1A; GRAF-1EF; HNGC:16303; LENG7; PTI1
UniProt ID: (Human) P68104
Entrez Gene ID: (Human) 1915
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.