Abnova
Manufacturer Code:H00001810B01
Catalog # 89-005-261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | DR1 (NP_001929.1 1 a.a. ~ 176 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human DR1 protein.
For Research Use Only
Protein Aliases: Down-regulator of transcription 1; down-regulator of transcription 1, TBP-binding (negative cofactor 2); NC2; NC2-BETA; negative cofactor 2; negative cofactor 2 beta; Negative cofactor 2-beta; Protein Dr1; TATA-binding protein-associated phosphoprotein
Gene Aliases: DR1; NC2; NC2-BETA; NC2B
UniProt ID: (Human) Q01658
Entrez Gene ID: (Human) 1810
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.