Abnova
Manufacturer Code:H00051181B02
Catalog # 89-007-609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | DCXR (NP_057370.1 1 a.a. ~ 244 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human DCXR protein.
For Research Use Only
Protein Aliases: carbonyl reductase 2; Carbonyl reductase II; DCR; Dicarbonyl/L-xylulose reductase; HCR2; HCRII; KIDCR; Kidney dicarbonyl reductase; L-xylulose reductase; P34H; SDR20C1; Short chain dehydrogenase/reductase family 20C member 1; short chain dehydrogenase/reductase family 20C, member 1; Sperm surface protein P34H; XR
Gene Aliases: DCR; DCXR; HCR2; HCRII; KIDCR; P34H; PNTSU; SDR20C1; XR
UniProt ID: (Human) Q7Z4W1
Entrez Gene ID: (Human) 51181
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.