Test
Invitrogen
Catalog # HYB 094-05-02
Quantity:
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export as a spreadsheet
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer. |
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com.
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase.
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | 1:8000 |
Host / Isotype | Mouse / IgG2b, kappa |
Class | Monoclonal |
Type | Antibody |
Clone | 1E7 |
Immunogen | Ovalbumin isolated from chicken egg white, denatured by heat and formaldehyde. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | 1 mg/mL |
Storage conditions | 4° C, store in dark |
RRID | AB_1072959 |
HYB 094-05-02 is specific for denatured chicken ovalbumin, but cross-reacts with native OA. Cross-reactivity to egg albumins from other species has not been tested.
HYB 094-05-02 has been successfully used in ELISA procedures. HYB 094-05-02 reacts with OA in denatured or modified forms (native OA surface-adsorbed, heat-denatured or reduced). Strong reaction is seen in ELISA, with OA directly coated onto the microtiter well.
The HYB 094-05-02 immunogen is OA isolated from chicken egg white, denatured by heat and formaldehyde.
Epitope specificity of HYB 094-05-02 differs from that of HYB 094-06-02 and HYB 094-07-02, as determined by inhibition ELISA.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Ovalbumin (OA), formaldehyde treated and heat denatured by incubation for 1 h at 105°C. OA is isolated from chicken egg white in which it makes up about 60% of solids. Native ovalbumin is a globular monomer of molecular mass ~45 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.