Abnova
Manufacturer Code:PAB12557
Catalog # 89-115-924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 700-796 of <i>Caenorhabditis elegans</i> daf-3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of daf-3.
For Research Use Only
Protein Aliases: Abnormal dauer formation protein 3; Smad protein; Smad protein daf-3
Gene Aliases: CELE_F25E2.5; daf-3; F25E2.5
UniProt ID: (C. elegans) Q95QI7
Entrez Gene ID: (C. elegans) 180431
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.