Proteintech
Manufacturer Code:163141AP150UL
Catalog # 50-173-0869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | dGK Fusion Protein Ag9361 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deoxyadenosine kinase, mitochondrial; Deoxyguanosine kinase dGK DGUOK; Deoxyguanosine kinase, mitochondrial
Gene Aliases: DGK; dGK1; dGK2; dGK3; DGUOK; MTDPS3
UniProt ID: (Human) Q16854, (Mouse) Q9QX60
Entrez Gene ID: (Human) 1716, (Mouse) 27369, (Rat) 297389
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.