Abnova
Manufacturer Code:H00120227B01
Catalog # 89-018-926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP2R1 (NP_078790.2 1 a.a. ~ 501 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human CYP2R1 protein.
For Research Use Only
Protein Aliases: Cytochrome P450 2R1; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; MGC4663; Vitamin D 25-hydroxylase
Gene Aliases: CYP2R1
UniProt ID: (Human) Q6VVX0
Entrez Gene ID: (Human) 120227
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.