Abnova
Manufacturer Code:H00000875A01
Catalog # 89-004-052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CBS (AAH11381.1 1 a.a. ~ 551 a.a) full-length recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant CBS.
For Research Use Only
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; Cystathionine beta-synthase-like protein; HIP4; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: CBS; CBSL; HIP4
UniProt ID: (Human) P35520
Entrez Gene ID: (Human) 875
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.