Proteintech
Manufacturer Code:163571AP150UL
Catalog # 50-173-0890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | cyclin I Fusion Protein Ag0493 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCNI CYC1 cyclin I CYI; cyclin ITI; Cyclin-I
Gene Aliases: CCNI; CCNI1; CYC1; CYI
UniProt ID: (Human) Q14094, (Mouse) Q9Z2V9
Entrez Gene ID: (Human) 10983, (Rat) 289500, (Mouse) 12453
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.