R&D Systems
Manufacturer Code:AF4079SP
Catalog # AF4079SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human CSL Gly208-Ser500 Accession # Q06330 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CBF-1; CBF1 CBF-1 csl H-2K binding factor-2 IGKJRB1 IGKJRB1RBPSUHCBF1 IGKJRBRBP-J kappa immunoglobulin kappa J region recombination signal binding protein 1 J kappa-recombination signal-binding protein KBF2 RBPJ RBP-JK RBPJKMGC61669 RBP-Jrecombining binding protein suppressor of hairless (Drosophila) RBPSUH recombination signal binding protein for immunoglobulin kappa J region recombining binding protein suppressor of hairless Renal carcinoma antigen NY-REN-30 SUH suppressor of hairless homolog; H-2K binding factor-2; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; RBP-J; RBP-J kappa; RBP-JK; Recombining binding protein suppressor of hairless; Renal carcinoma antigen NY-REN-30; suppressor of hairless homolog
Gene Aliases: AOS3; CBF1; csl; IGKJRB; IGKJRB1; KBF2; RBP-J; RBPJ; RBPJK; RBPSUH; SUH
UniProt ID: (Human) Q06330
Entrez Gene ID: (Human) 3516
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.