Abnova
Manufacturer Code:H00056913M01
Catalog # 89-014-322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F1 |
Immunogen | C1GALT1 (NP_064541 264 a.a. ~ 363 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant C1GALT1.
For Research Use Only
Protein Aliases: B3Gal-T8; Beta-1,3-galactosyltransferase; C1GALT; C1GalT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T1; Core 1 O-glycan T-synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; Core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; Glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; T-synthase
Gene Aliases: C1GALT; C1GALT1; T-synthase
UniProt ID: (Human) Q9NS00
Entrez Gene ID: (Human) 56913
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.